Learn More
Medchemexpress LLC Polyvinyl alcohol | 9002-89-5 | MFCD00081922 | 99.0% | 146,000-186,000 g/mol | (C2H4O)n | 25g
SDP
Supplier:  Medchemexpress LLC HYY085025G
Encompass_Preferred
Polyvinyl alcohol (Mw 146000-186000 99 % hydrolyzed) is a biodegradable semi-crystalline synthetic polymer with a high degree of biocompatibility in vitro and in vivo Polyvinyl alcohol can be used in the study of drug delivery system in cancer[1][2]
Catalog No. 50-002-68676