Learn More
Medchemexpress LLC Polyvinyl alcohol | 9002-89-5 | MFCD00081922 | >99.0% | 146000-186000 | [-CH2CH(OH)-]n | 10g
SDP
Supplier:  Medchemexpress LLC HYY085010G
Encompass_Preferred
Polyvinyl alcohol (Mw 146000-186000 99 % hydrolyzed) is a biodegradable semi-crystalline synthetic polymer with a high degree of biocompatibility in vitro and in vivo Polyvinyl alcohol can be used in the study of drug delivery system in cancer[1][2]
Catalog No. 50-002-69754