Learn More
Medchemexpress LLC Polyvinyl alcohol | 9002-89-5 | MFCD00677796 | 98.0% | 13,000-23,000 g/mol | C2H4O | 10g
SDP
Supplier:  Medchemexpress LLC HYY0850N10G
Encompass_Preferred
Polyvinyl alcohol (Mw 13000-23000 98% hydrolyzed) also known as PVA is a vinyl water-soluble polymer that can be used as a non-ionic surfactant It can also be used as a biodegradable polymer and can be used in adhesives coatings textiles ceramics and cosmetics[1]
Catalog No. 50-002-69907