Learn More
Medchemexpress LLC Polyvinyl alcohol (Mw 146000-186000, 87-89% hydrolyzed) | 9002-89-5 | MFCD00081922 | 88.1% | 146,000-186,000 | (C2H4O)x | 50g
SDP
Supplier:  Medchemexpress LLC HYY085050G
Encompass_Preferred
Polyvinyl alcohol (Mw 146000-186000 99 % hydrolyzed) is a biodegradable semi-crystalline synthetic polymer with a high degree of biocompatibility in vitro and in vivo Polyvinyl alcohol can be used in the study of drug delivery system in cancer[1][2]
Catalog No. 50-002-69755