Learn More
Medchemexpress LLC Polyvinyl alcohol (Mw 89,000-98,000, 99+% hydrolyzed) | 9002-89-5 | 99.5% | 89,000-98,000 (approx.) | (C2H4O)x | 5g
SDP
Supplier:  Medchemexpress LLC HYY0850H5G
Encompass_Preferred
Polyvinyl alcohol (Mw 89000-98000 99 % hydrolyzed) also known as PVA is a vinyl water-soluble polymer that can be used as a non-ionic surfactant It can also be used as a biodegradable polymer and can be used in adhesives coatings textiles ceramics and cosmetics[1]
Catalog No. 50-002-68675