Learn More
Sigma Aldrich Fine Chemicals Biosciences Polyvinylpyrrolidone, 9003-39-8, MFCD00149016, 100 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences PVP40100G
Encompass
Linear Formula: (C6H9NO)n, average mol wt 40,000, Synonym: PVP, Polyvidone, Povidone. Polyvinylpyrrolidone has been used in the purification of glutamine synthetases from plant samples. It has been used to block the PVDF (polyvinylidene difluoride) membrane and to avoid nonspecific binding.
Catalog No. NC1230803