Learn More
Medchemexpress LLC Polyvinylpyrrolidone, average Mw~29000 | 9003-39-8 | 10 G
SDP
Supplier:  Medchemexpress LLC HYB1620K10G
Encompass_Preferred
Polyvinylpyrrolidone, average Mw~29000 is a multifunctional synthetic polymer with an average molecular weight of 29,000 Da. It serves as a versatile excipient in both conventional formulations and novel controlled or targeted delivery systems.
  • Functions as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.
  • Shows affinity and adsorption for Indomethacin.
  • Inhibits drug crystallization and promotes amorphous state.
Catalog No. 50-004-13260