Learn More
Medchemexpress LLC Polyvinylpyrrolidone, average Mw 360000 | 9003-39-8 | MFCD01076626 | 100.0% | 500 G
SDP
Supplier:  Medchemexpress LLC HYB1620N500G
Encompass_Preferred
Polyvinylpyrrolidone, average Mw 360000, is a multifunctional synthetic polymer with an average molecular weight of 360000 Da. It is widely used to synthesize nanoparticles and is a versatile excipient for both conventional formulations and novel controlled or targeted delivery systems. In pharmacy, its inertness, non-toxicity, and biocompatibility make it suitable for various applications.
  • Functions as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer
  • Improves drug bioavailability and stability
  • Enhances physicomechanical properties of preparations
  • Adjusts drug release rates
  • Prolongs the in vivo circulation time of liposomes
Catalog No. 50-004-13128