Learn More
Medchemexpress LLC Polyvinylpyrrolidone K15 | 9003-39-8 | 98.0% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYB1620B
Encompass_Preferred
Polyvinylpyrrolidone K15 is a linear non-ionic polymer that creates vigorous bright crystalline layers and possesses adhesive, cohesive, and dispersive properties. It is widely utilized in various fields including paper, fibers, textiles, ceramics, and pharmaceutics. Its inertness, non-toxicity, and biocompatibility make it a versatile excipient for drug delivery systems, serving as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.
  • Creates vigorous bright crystalline layers
  • Possesses adhesive, cohesive, and dispersive properties
  • Utilized in paper, fibers, textiles, ceramics, and pharmaceutics
  • Inert and non-toxic
  • Biocompatible
  • Functions as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer
Catalog No. 50-004-14268