Learn More
Medchemexpress LLC Polyvinylpyrrolidone K90 | 9003-39-8 | 98.0% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB1620H10G
Encompass_Preferred
Polyvinylpyrrolidone K90 (PVP K90) is a versatile polymer used in various applications, including drug delivery systems. It is effective as a wound healing promoter and disinfectant when combined with iodine and other compounds. PVP K90 also stabilizes nanosuspensions and interacts with the surface of drug crystals.
  • Can be used to prepare electrospun nanofibrous membranes
  • Effective wound healing promoter
  • Disinfectant when combined with iodine and other compounds
  • Stabilizes nanosuspensions
  • Interacts with the surface of drug crystals
Catalog No. 50-004-13259