Learn More
Cayman Chemical PomalIdomIde 4-PEG5-amInh 25mg
SDP
Supplier:  Cayman Chemical 3998725 MG
Encompass_Preferred
A PEGylated form of pomalidomide; has been conjugated to various ligands in the development of PROTACs
Catalog No. 50-294-2080