Learn More
Medchemexpress LLC Pomalidomide-5-C9-NH2 (hydrochloride) | 2925306-22-3 | 98.7% | 450.96 g/mol | C22H31ClN4O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15526510MG
Encompass_Preferred
Pomalidomide-5-C9-NH2 hydrochloride is a pomalidomide-derived ligand for the cereblon (CRBN) E3 ubiquitin ligase, supplied as the hydrochloride salt for research use. It is primarily used as a CRBN-recruiting moiety in the design and synthesis of PROTACs and other targeted protein degradation tools, and is provided in small milligram quantities with high reported purity.
  • Ligand for cereblon (CRBN) E3 ubiquitin ligase.
  • Designed for use as a CRBN-recruiting moiety in PROTAC synthesis.
  • Provided as the hydrochloride salt for improved stability and handling.
  • High reported purity (98.68%) suitable for research applications.
  • Available in small milligram quantities for medicinal chemistry workflows.
Catalog No. 50-002-87468