Learn More
Medchemexpress LLC Pomalidomide-5'-PEG5-C2-COOH | 2412056-50-7 | 98.1% | 565.57 | 50 MG

Supplier: Medchemexpress LLC HY14848450MG
Pomalidomide-5'-PEG5-C2-COOH is an active compound used for the research of various biochemical applications.
- Active compound
- Can be used for research of various biochemical applications
- Store at -20°C, protected from light, and under nitrogen
- Soluble in DMSO
- Molecular formula C26H35N3O11
- Purity of 98.1%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.