Learn More
Medchemexpress LLC Pomalidomide-amido-C1-Br | 2351106-38-0 | MFCD32903493 | >98.0% | C15H12BrN3O5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13061710MG
Encompass_Preferred
Pomalidomide-amido-C1-Br is an E3 ligase ligand-linker conjugate that incorporates a pomalidomide-based cereblon ligand for use in targeted protein degradation and PROTAC research. The compound is supplied as a high-purity research reagent (CAS 2351106-38-0) with molecular formula C15H12BrN3O5 and molecular weight 394.18 g·mol⁻¹. It is typically provided in milligram-scale quantities for screening and synthetic applications.
  • High purity suitable for biochemical research.
  • Contains a pomalidomide-based cereblon ligand for PROTAC design.
  • Provided in small milligram-scale pack sizes for screening and synthesis.
  • Characterized by molecular formula and molecular weight for identity confirmation.
  • Supports design and development of E3 ligase recruiting degraders.
Catalog No. 50-002-60030