Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-amido-C5-PEG2-C6-chlorine | 1835705-69-5 | 98.75% | 594.10 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1379051MG

Encompass_Preferred

Pomalidomide-amido-C5-PEG2-C6-chlorine is a derivative of Pomalidomide (HY-10984). It is supplied as a solid, white to off-white in color, and is intended for research use only, not for sale to patients.

  • Derivative of Pomalidomide
  • For research use only
  • Solid, white to off-white appearance
  • Ships at room temperature in continental US
  • Store at 4°C under nitrogen
  • Store in solvent at -80°C for 6 months
  • Store in solvent at -20°C for 1 month

Catalog No. 50-003-52237


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.