Learn More
Medchemexpress LLC Pomalidomide-amino-PEG5-NH2 hydrochloride | 2421217-05-0 | MFCD01304495 | 99.6% | C25H35ClN4O10 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13381610MG
Encompass_Preferred
Pomalidomide-amino-PEG5-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate combining a pomalidomide-derived cereblon ligand with a PEG5 amine linker. Supplied as the hydrochloride salt, it is intended as a research building block for proteolysis targeting chimera (PROTAC) design and ligand-linker conjugation studies.
  • Suitable for PROTAC synthesis and E3 ligase targeting.
  • Hydrochloride salt improves handling and stability.
  • High purity appropriate for research applications.
  • Soluble in DMSO at high concentration; ultrasonic assistance recommended.
  • Stable when stored sealed at 4°C; defined solvent storage conditions extend shelf life.
  • Supplied as a solid for convenient weighing and storage.
Catalog No. 50-002-61861