Learn More
Medchemexpress LLC Pomalidomide-C2-amido-(C1-O-C5-O-C1)2-COOH | 2351103-63-2 | 93.9% | 100 MG

Supplier: Medchemexpress LLC HY128848100MG
Pomalidomide-C2-amido-(C1-O-C5-O-C1)2-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and a linker used in PROTAC technology. This product is for research use only.
- Incorporates Pomalidomide based cereblon ligand
- Used in PROTAC technology
- Targets cereblon
- Molecular weight: 648.70
- Formula: C31H44N4O11
- Appearance: Oil
- Color: Light brown to brown
- Purity: 93.92%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.