Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-C3-NHS ester | 2828476-87-3 | 98.7% | 456.41 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1481425MG

Encompass_Preferred

Pomalidomide-C3-NHS ester is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Pomalidomide based cereblon ligand and an NHS ester linker. This product is for research use only and is not sold to patients.

  • E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Features an NHS ester linker
  • Target: Cereblon
  • Appearance: Solid, light yellow to yellow
  • Soluble in DMSO

Catalog No. 50-003-60636


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.