Learn More
Medchemexpress LLC POMALIDOMIDE-C7-COOH - 100MG
SDP
Supplier:  Medchemexpress LLC HY133799100MG
Encompass_Preferred
Pomalidomide-C7-COOH is a synthesized E3 ligase cereblon ligand-linker conjugate. Pomalidomide-C7-COOH is an intermediate for the synthesis of PROTAC BCL-XL degraders.Purity: 98.04%.CAS No. : 2225940-51-0
Catalog No. NC1904255