Learn More
Selleck Chemical LLC Pomalidomide 50mg 19171-19-8 CC-4047
SDP
Supplier:  Selleck Chemical LLC S156750MG
Encompass_Preferred
Pomalidomide (CC-4047) inhibits LPS-induced TNF-? release with IC50 of 13 nM in PBMCs. Pomalidomide can be utilized in PROTAC as a ligand for targeting E3 ligase and inhibiting the E3 ligase protein cereblon (CRBN). Pomalidomide promotes apoptosis and cell cycle arrest. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-734-7