Learn More
Medchemexpress LLC Pomalidomide-CO-C5-Br | 2227423-38-1 | 450.28 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY169010100MG
Encompass_Preferred
Pomalidomide-CO-C5-Br (Compound 6b) is a conjugated compound of the E3 ligand Pomalidomide and a linker. It can be used to synthesize highly effective PROTAC molecules targeting ADC. It is intended for research use only and not for sale to patients.
  • Conjugated compound of E3 ligand Pomalidomide and linker.
  • Used to synthesize PROTAC molecules.
  • Targets ADCs.
  • For research use only.
Catalog No. 50-003-90453