Learn More
Medchemexpress LLC Pomalidomide-CO-C5-Br | 2227423-38-1 | 450.28 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1690101G
Encompass_Preferred
Pomalidomide-CO-C5-Br (Compound 6b) is a conjugated compound of the E3 ligand Pomalidomide and linker. It is designed for use in synthesizing highly effective PROTAC molecules.
  • Conjugated compound of E3 ligand Pomalidomide and linker.
  • Used to synthesize highly effective PROTAC molecules.
  • Targets ADC.
Catalog No. 50-003-90467