Learn More
Cayman Chemical PomalIdomIde-d5 1mg
SDP
Supplier:  Cayman Chemical 253581 MG
Encompass_Preferred
An internal standard for the quantification of pomalidomide by GC- or LC-MS
Catalog No. 50-293-3453