Learn More
TARGETMOL CHEMICALS INC POMALIDOMIDE-PEG1-CO2H 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T4002425MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Pomalidomide-PEG1-CO2H is a synthetic E3 ligase ligand-linker conjugate containing a Pomalidomide-based cereblon ligand and a 2-unit PEG linker. purity: 97%
Catalog No. 50-257-6833