Learn More
Medchemexpress LLC Pomalidomide-PEG2-acetic acid | 2143097-05-4 | 99.4% | 419.39 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1677595MG
Encompass_Preferred
Pomalidomide-PEG2-acetic acid is a chemical conjugate designed for specific biological applications. It functions as an E3 ligase ligand-linker conjugate and is primarily utilized for PROTAC synthesis. This product is intended for research purposes only and is not suitable for medical applications or sale to patients.
  • Conjugate composed of an E3 ligase ligand and a linker
  • Used for PROTAC synthesis
  • For research use only
Catalog No. 50-003-93494