Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-PEG2-acetic acid | 2143097-05-4 | 99.4% | 419.39 g·mol⁻1 | C19H21N3O8 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY16775910MG

Encompass_Preferred

Pomalidomide-PEG2-acetic acid is a pomalidomide-derived cereblon E3 ligase ligand-linker conjugate that incorporates a short PEG2 spacer terminated with an acetic acid (-CH2COOH) functional group for conjugation. It is supplied as a solid building block intended for PROTAC synthesis and related coupling chemistries.

  • Used as an E3 ligase ligand-linker building block for PROTAC synthesis.
  • PEG2 linker with a terminal acetic acid group for amide or ester coupling.
  • High purity (99.43%).
  • Molecular formula C19H21N3O8 and molecular weight 419.39 g·mol⁻1.
  • CAS number 2143097-05-4.
  • Available in mg-scale quantities for synthetic applications.

Catalog No. 50-002-89407


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.