Learn More
Medchemexpress LLC Pomalidomide-PEG2-azide | 2267306-14-7 | 96.6% | 444.40 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY137537
Encompass_Preferred
This product is an E3 ligase ligand-linker conjugate incorporating a cereblon ligand and a 2-unit PEG linker. It functions as a click chemistry reagent, featuring an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition reactions with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition reactions with molecules containing DBCO or BCN groups.
  • Synthesized E3 ligase ligand-linker conjugate.
  • Includes a cereblon ligand.
  • Features a 2-unit PEG linker.
  • Functions as a click chemistry reagent.
  • Contains an Azide group.
  • Facilitates copper-catalyzed azide-alkyne cycloaddition.
  • Supports strain-promoted alkyne-azide cycloaddition.
Catalog No. 50-003-50693