Learn More
Medchemexpress LLC HY-137537 50mg Medchemexpress, Pomalidomide-PEG2-azide CAS:2267306-14-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13753750MG
Encompass_Preferred
Medchemexpress, HY-137537 50mg Pomalidomide-PEG2-azide CAS:2267306-14-7 Pomalidomide-PEG2-azide is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and 2-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1497