Learn More
Medchemexpress LLC Pomalidomide-PEG4-azide | 2271036-47-4 | 99.85% | C23H30N6O8 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14101550MG
Encompass_Preferred
Pomalidomide-PEG4-azide is a synthesized E3 ligase ligand-linker conjugate designed for PROTAC technology. It incorporates a Pomalidomide-based cereblon ligand and functions as a click chemistry reagent. This compound contains an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates cereblon ligand
  • Used in PROTAC technology
  • Click chemistry reagent with Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Targets Cereblon
Catalog No. 50-003-54973