Learn More
TARGETMOL CHEMICALS INC Pomalidomide-PEG4-C-COOH 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1732510MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Pomalidomide-PEG4-C-COOH (E3 Ligase Ligand-Linker Conjugates 1) is a synthesized E3 ligase ligand-linker conjugate that combines the Pomalidomide-based cereblon ligand and 4-unit PEG linker used in PROTAC technology. Purity 96%
Catalog No. 50-287-8283