Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-PEG4-C-COOH | 2097938-44-6 | MFCD31540390 | 99.2% | 507.49 g·mol⁻¹ | C23H29N3O10 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY2193010MG

Encompass_Preferred

Pomalidomide-PEG4-C-COOH is an E3 ligase ligand-linker conjugate that combines a pomalidomide-derived cereblon ligand with a four-unit polyethylene glycol (PEG4) spacer terminating in a carboxylic acid. It is intended for use in targeted protein degradation research and PROTAC design, providing a conjugation handle and improved linker flexibility and solubility.

  • Contains pomalidomide-based cereblon ligand.
  • Four-unit PEG linker provides flexibility and solubility.
  • Terminal carboxylic acid enables conjugation to target ligands.
  • Designed for PROTAC design and chemical biology applications.
  • High purity (99.19% by LCMS).
  • Molecular weight 507.49 g·mol⁻¹; formula C23H29N3O10.
  • Available in small research quantities suitable for assay development.

Catalog No. 50-002-97767


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.