Learn More
Medchemexpress LLC Pomalidomide-PEG4-C2-NH2 | 2225940-52-1 | 95.1% | 492.52 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1125991G
Encompass_Preferred
Pomalidomide-PEG4-C2-NH2 is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and a 4-unit PEG linker, primarily utilized in PROTAC technology.
  • Used in PROTAC technology
  • Incorporates Pomalidomide based cereblon ligand
  • Features a 4-unit PEG linker
  • Target: E3 ligase ligand-linker conjugates
Catalog No. 50-004-37574