Learn More
Medchemexpress LLC Pomalidomide-PEG4-C2-NH2 hydrochloride | 2357105-92-9 | 98.2% | 528.98 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY112599B100MG
Encompass_Preferred
Pomalidomide-PEG4-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Pomalidomide-based cereblon ligand and a 4-unit PEG linker, which are utilized in the synthesis of PROTACs. PROTACs themselves consist of two different ligands linked together, one targeting an E3 ubiquitin ligase and the other a protein of interest. Their function is to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide-based cereblon ligand
  • Contains a 4-unit PEG linker
  • Used in the synthesis of PROTACs
  • For research use only
Catalog No. 50-004-36189