Learn More
Medchemexpress LLC Pomalidomide-PEG4-C2-NH2 hydrochloride | 2357105-92-9 | 98.2% | 528.98 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY112599B
Encompass_Preferred
Pomalidomide-PEG4-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Pomalidomide-based cereblon ligand and a 4-unit PEG linker, used in the synthesis of PROTACs. PROTACs utilize an E3 ubiquitin ligase ligand and a target protein ligand, exploiting the ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide-based cereblon ligand
  • Features a 4-unit PEG linker
  • Used in the synthesis of PROTACs
  • Targets cereblon
  • For research use only
Catalog No. 50-004-39529