Learn More
Medchemexpress LLC Pomalidomide-PEG4-C2-NH2 hydrochloride | 2357105-92-9 | 98.2% | 528.98 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY112599B50MG
Encompass_Preferred
Pomalidomide-PEG4-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and 4-unit PEG linker used in the synthesis of PROTACs. It is for research use only and not sold to patients.
  • E3 ligase ligand-linker conjugate
  • Incorporates pomalidomide based cereblon ligand and 4-unit PEG linker
  • Used in the synthesis of PROTACs
  • Appearance: Solid-liquid mixture
  • Color: Light yellow to yellow
Catalog No. 50-004-37684