Learn More
Medchemexpress LLC Pomalidomide-PEG6-butyl iodide | 1835705-74-2 | 98.4% | 761.60 | 25 MG

Supplier: Medchemexpress LLC 25MGHY136156
Pomalidomide-PEG6-butyl iodide is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and a 6-unit PEG linker used in PROTAC technology. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
- Incorporates Pomalidomide based cereblon ligand
- Features a 6-unit PEG linker
- Used in PROTAC technology
- Exploits the intracellular ubiquitin-proteasome system
- For research use only
- Target: Cereblon
- Appearance: White to off-white solid
- Purity: 98.35%
- Molecular formula: C31H44IN3O11
- Molecular weight: 761.60
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.