Learn More
Medchemexpress LLC Pomalidomide-PEG6-C2-COOH | 2225148-49-0 | 98.95% | C28H39N3O12 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY141658
Encompass_Preferred
Pomalidomide-PEG6-C2-COOH is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Pomalidomide-based cereblon ligand and a 6-unit PEG linker, utilized in PROTAC technology.
  • Synthesized E3 ligase ligand-linker conjugate.
  • Incorporates a Pomalidomide-based cereblon ligand.
  • Features a 6-unit PEG linker.
  • Utilized in PROTAC technology.
Catalog No. 50-003-55920