Learn More
Medchemexpress LLC Pomalidomide-PEG6-C2-COOH | 2225148-49-0 | 99.0% | C28H39N3O12 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141658100MG
Encompass_Preferred
Pomalidomide-PEG6-C2-COOH is a synthesized E3 ligase ligand-linker conjugate. It incorporates a cereblon ligand based on Pomalidomide and a 6-unit PEG linker, making it suitable for use in PROTAC technology. This product is intended for research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a cereblon ligand
  • Contains a 6-unit PEG linker
  • Used in technology for targeted protein degradation
Catalog No. 50-003-53738