Learn More
Medchemexpress LLC Pomalidomide-PEG6-C2-COOH | 2225148-49-0 | 99.0% | C28H39N3O12 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY141658
Encompass_Preferred
Pomalidomide-PEG6-C2-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide-based cereblon ligand and a 6-unit PEG linker used in PROTAC technology. It has a molecular weight of 609.62 and appears as a viscous liquid, light yellow to yellow in color. This product is for research use only.
  • E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Contains a 6-unit PEG linker
  • Used in PROTAC technology
Catalog No. 50-003-52502