Learn More
Medchemexpress LLC Pomalidomide-PEG6-C2-COOH | 2225148-49-0 | MFCD32173791 | 98.0% | 609.6 g/mol | C28H39N3O12 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1416585MG
Encompass_Preferred
Pomalidomide-PEG6-C2-COOH is an E3 ligase ligand-linker conjugate intended for PROTAC synthesis and chemical biology research. It couples a pomalidomide-derived cereblon-binding moiety to a six-unit polyethylene glycol spacer that terminates in a C2 carboxylic acid, providing a functional handle for conjugation while improving solubility and linker flexibility.
  • Facilitates cereblon-mediated target recruitment via a pomalidomide-based ligand.
  • Peg6 spacer provides flexibility and improved solubility.
  • Terminal carboxylic acid enables straightforward coupling chemistry.
  • Suitable for PROTAC design and medicinal chemistry applications.
  • Provided as a high-purity reagent for research use.
Catalog No. 50-002-21351