Learn More
Medchemexpress LLC Pomalidomide-PEG6-NH2 hydrochloride | 2341841-01-6 | MFCD32734576 | 98.0% | C25H36ClN3O10 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13616110MG
Encompass_Preferred
Pomalidomide-PEG6-NH2 hydrochloride is an E3 ligase ligand-linker conjugate that couples a pomalidomide-based cereblon ligand to a six-unit polyethylene glycol (PEG6) spacer. Supplied as the hydrochloride salt, it is intended for research use in PROTAC design and chemical biology applications; molecular formula C25H36ClN3O10, molecular weight 574.02 g·mol⁻¹.
  • Provides a pomalidomide-based cereblon binding moiety.
  • Includes a PEG6 linker to improve solubility and flexibility.
  • Hydrochloride salt form enhances stability and handling.
  • High purity suitable for PROTAC synthesis and analytical work.
  • Well characterized with supporting COA and datasheet documentation.
Catalog No. 50-002-59899