Learn More
Medchemexpress LLC Ponasterone A | 13408-56-5 | 464.63 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN15345MG
Encompass_Preferred
Ponasterone A (25-Deoxyecdysterone) is an ecdysteroid with strong affinity for the ecdysone receptor. It is a potent regulator of gene expression in cells and transgenic animals, enabling reporter genes to be turned on and off rapidly.
  • Ecdysteroid
  • Strong affinity for the ecdysone receptor
  • Potent regulator of gene expression
  • Enables reporter genes to be turned on and off rapidly
Catalog No. 50-004-16515