Learn More
Apexbio Technology LLC Ponceau 4R 2611-82-7 1g
SDP
Supplier:  Apexbio Technology LLC A47271000
Encompass_Preferred
Ponceau 4R (CAS 2611-82-7) is a synthetic azo dye widely used as a chromogenic agent in staining applications Ponceau 4R is valued for its chemical stability under varied experimental conditions including heat acidic pH and light exposure It facilitates visualization protocols in studies involving dye-protein interactions dye stability and metabolic kinetics Based on these properties Ponceau 4R holds research potential in molecular biology bioanalytical experiments and food chemistry
Catalog No. 50-000-95101