Learn More
Medchemexpress LLC Ponceau 4R | 2611-82-7 | 85.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYD0193A100MG
Encompass_Preferred
Ponceau 4R (85%), also known as Acid Red 18 (85%) or New Coccine (85%), is a synthetic colorant commonly used as a food coloring. This strawberry red azo dye is typically synthesized from aromatic hydrocarbons. It demonstrates stability to light, heat, and acid, but its color can fade in the presence of ascorbic acid.
  • Synthetic colorant for various applications
  • Strawberry red azo dye
  • Stable to light, heat, and acid
  • Fades in the presence of ascorbic acid
  • For research use only
Catalog No. 50-004-10193