Learn More
Medchemexpress LLC Ponceau 4R | 2611-82-7 | 99.8% | 5 G
SDP
Supplier:  Medchemexpress LLC HYD01935G
Encompass_Preferred
Ponceau 4R is a synthetic colorant commonly used for food coloring. This strawberry red azo dye is typically synthesized from aromatic hydrocarbons and exhibits stability to light, heat, and acids. It can be utilized in various food applications. For research use only.
  • Can be used for food coloring
  • Strawberry red azo dye
  • Synthesized from aromatic hydrocarbons
  • Stable to light, heat, and acids
  • Suitable for various food applications
Catalog No. 50-004-10270