Learn More
Medchemexpress LLC Ponceau 4R | 2611-82-7 | 99.8% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYD0193500MG
Encompass_Preferred
Ponceau 4R is a synthetic colorant, also known as Acid Red 18 or New Coccine, commonly used as a food coloring. This strawberry red azo dye is synthesized from aromatic hydrocarbons and exhibits stability to light, heat, and acids. It is intended for research use only, serving as a laboratory chemical for the manufacture of substances.
  • Stable to light, heat, and acids
  • Suitable for research use only
  • Used in laboratory chemical applications
  • Strawberry red azo dye
Catalog No. 50-004-09938