Learn More
Medchemexpress LLC Poncirin | 14941-08-3 | MFCD23380546 | 10 MG

Supplier: Medchemexpress LLC HYN225810MG
Poncirin is a flavonoid glycoside used as a research reagent and analytical standard in studies of inflammation and pain modulation.
- Flavonoid glycoside (isosakuranetin 7-O-neohesperidoside).
- Molecular formula C28H34O14; molecular weight 594.56 g/mol.
- CAS number 14941-08-3.
- Intended for research use; not for human or clinical use.
- Storage: 4°C, protect from light; in solvent, -80°C for 6 months or -20°C for 1 month.
- Common applications include anti-inflammatory and anti-hyperalgesia research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.