Learn More
TARGETMOL CHEMICALS INC PONESIMOD 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T325810MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ponesimod (ACT-128800) is an orally available sphingosine-1-phosphate receptor 1 (S1PR1 S1P1) agonist with potential immunomodulating activity. purity: 99%
Catalog No. 50-257-6967