Learn More
Medchemexpress LLC HY-10569 10mg , Ponesimod CAS:854107-55-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1056910MG
Encompass_Preferred
HY-10569 10mg Ponesimod CAS: 854107-55-4 Ponesimod (ACT-128800) is a potent, selective and orally active agonist of S1P1, with an IC50 of 6 nM in a radioligand binding assay. Ponesimod activates S1P1-mediated signal transduction with high potency (EC50=5.7 nM). Ponesimod can protect against lymphocyte-mediated tissue inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0475