Learn More
TARGETMOL CHEMICALS INC PONICIDIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2S19755MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ponicidin (Rubescensine B) is a diterpenoid derived from Rabdosia rubescens. Ponicidin exhibits immunoregulatory anti-inflammatory anti-viral and anti-cancer activity. Ponicidin induces apoptosis of gastric carcinoma cells decreases the phosphorylation of JAK2 and STAT3 and shows no effect on protein levels of JAK2 and STAT3. purity: 99%
Catalog No. 50-258-0436